Assay Details
Target Gene Details
Entrez Gene ID: | 3556 |
Gene Name: | interleukin 1 receptor accessory protein |
Gene Aliases: |
C3orf13, IL-1RAcP, IL1R3 |
Location: |
Chr.3:190514051-190657197 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RAP | NM_001167928.1 | NP_001161400.1 | ||
| NM_001167929.1 | NP_001161401.1 | |||
| NM_001167930.1 | NP_001161402.1 | |||
| NM_001167931.1 | NP_001161403.1 | |||
| NM_002182.3 | NP_002173.1 | |||
| NM_134470.3 | NP_608273.1 | |||
| XM_017006347.1 | XP_016861836.1 | |||
| AB006537.1 | BAA25421.1 | |||
| AF538730.1 | AAQ01755.1 | |||
| AF538731.1 | AAQ01756.1 | |||
| AF538732.1 | AAQ01757.1 | |||
| AF538733.1 | AAQ01758.1 | |||
| AF538734.1 | AAQ01759.1 | |||
| AK291669.1 | ||||
| BC053621.1 | AAH53621.1 | |||
| DA024801.1 | ||||
| EF591790.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829820 | Chr.3:190379481 - 190544546 on Build GRCh38 | Gain |
|
| esv3893843 | Chr.3:190075998 - 191094501 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map