Assay Details
Target Gene Details
Entrez Gene ID: | 6480 |
Gene Name: | ST6 beta-galactoside alpha-2,6-sialyltransferase 1 |
Gene Aliases: |
SIAT1, ST6GalI, ST6N |
Location: |
Chr.3:186930343-187078553 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GAL1 | NM_173216.2 | NP_775323.1 | ||
| NM_173217.2 | NP_775324.1 | |||
| XM_006713734.1 | XP_006713797.1 | |||
| XM_011513085.1 | XP_011511387.1 | |||
| XM_011513086.1 | XP_011511388.1 | |||
| XM_017007065.1 | XP_016862554.1 | |||
| A17362.1 | CAA01327.1 | |||
| DB152155.1 | ||||
| DB235666.1 | ||||
| X17247.1 | CAA35111.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001111 | Chr.3:186796249 - 187014891 on Build GRCh38 | Gain |
|
| esv34136 | Chr.3:186697095 - 187036942 on Build GRCh38 | Loss |
|
| nsv508975 | Chr.3:186936621 - 186986854 on Build GRCh38 | Insertion |
|
| nsv1004008 | Chr.3:186879814 - 187204053 on Build GRCh38 | Gain |
|
| esv3569168 | Chr.3:186977101 - 186992817 on Build GRCh38 | Loss |
|
| esv2759207 | Chr.3:186891945 - 187066496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map