Assay Details
Target Gene Details
Entrez Gene ID: | 132203 |
Gene Name: | sentan, cilia apical structure protein |
Gene Aliases: |
S100A1L, S100AL, sentan |
Location: |
Chr.3:63652668-63665217 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNTN | NM_001080537.1 | NP_001074006.1 | ||
| AK126350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3849 | Chr.3:63632791 - 63666859 on Build GRCh38 | Insertion |
|
| nsv1008751 | Chr.3:63595517 - 63862172 on Build GRCh38 | Gain |
|
| nsv834716 | Chr.3:63577882 - 63787971 on Build GRCh38 | Loss |
|
| esv2422509 | Chr.3:63515793 - 63664263 on Build GRCh38 | Deletion |
|
| nsv460581 | Chr.3:63654925 - 63687658 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map