Assay Details
Target Gene Details
Entrez Gene ID: | 6565 |
Gene Name: | solute carrier family 15 member 2 |
Gene Aliases: |
PEPT2 |
Location: |
Chr.3:121894324-121944187 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC15A2 | NM_001145998.1 | NP_001139470.1 | ||
| NM_021082.3 | NP_066568.3 | |||
| XM_005247722.3 | XP_005247779.1 | |||
| XM_006713736.3 | XP_006713799.1 | |||
| XM_017007074.1 | XP_016862563.1 | |||
| AK289820.1 | ||||
| AK296895.1 | ||||
| AK302002.1 | ||||
| AK304189.1 | ||||
| BC020743.1 | ||||
| BC044572.1 | AAH44572.1 | |||
| S78203.1 | AAB34388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516233 | Chr.3:121700454 - 121951558 on Build GRCh38 | Gain+Loss |
|
| esv33139 | Chr.3:121781553 - 122372984 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map