Assay Details
Target Gene Details
Entrez Gene ID: | 25852 |
Gene Name: | armadillo repeat containing 8 |
Gene Aliases: |
GID5, HSPC056, S863-2, VID28 |
Location: |
Chr.3:138187248-138298389 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597911 | Chr.3:138130790 - 138220069 on Build GRCh38 | Gain |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| esv3597913 | Chr.3:138161103 - 138213569 on Build GRCh38 | Gain |
|
| nsv998106 | Chr.3:138132356 - 138215768 on Build GRCh38 | Gain |
|
| esv2759180 | Chr.3:137892947 - 138214993 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map