Assay Details
Target Gene Details
Entrez Gene ID: | 116933 |
Gene Name: | CLRN1 antisense RNA 1 |
Gene Aliases: |
CLRN1OS, UCRP |
Location: |
Chr.3:150972678-151079830 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLRN1-AS1 | NR_024066.1 | |||
| AF388367.1 | ||||
| AK126442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829756 | Chr.3:150974849 - 151171951 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111917373] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map