Assay Details
Target Gene Details
Entrez Gene ID: | 93624 |
Gene Name: | transcriptional adaptor 2B |
Gene Aliases: |
ADA2(beta), ADA2B |
Location: |
Chr.4:7043429-7057952 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TADA2B | NM_152293.2 | NP_689506.2 | ||
| XM_011513595.1 | 1 | 869 | XP_011511897.1 | |
| AK093974.1 | ||||
| AK096655.1 | 1 | 446 | ||
| AK126966.1 | ||||
| BC047794.1 | AAH47794.1 | |||
| BC101334.1 | ||||
| BC101335.1 | ||||
| BC101336.1 | ||||
| BC101337.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428436 | Chr.4:6986421 - 7268174 on Build GRCh38 | Gain |
|
| nsv4217 | Chr.4:7033785 - 7078503 on Build GRCh38 | Deletion |
|
| esv34033 | Chr.4:6971543 - 7388894 on Build GRCh38 | Loss |
|
| nsv469847 | Chr.4:7001798 - 7166714 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map