Assay Details
Target Gene Details
Entrez Gene ID: | 646603 |
Gene Name: | chromosome 4 open reading frame 51 |
Gene Aliases: |
- |
Location: |
Chr.4:145680103-145792248 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4orf51 | NM_001080531.1 | 1 | 120 | NP_001074000.1 |
| XM_006714289.3 | 1 | 221 | XP_006714352.1 | |
| XM_011532196.2 | 1 | 221 | XP_011530498.1 | |
| XM_011532197.2 | 1 | 221 | XP_011530499.1 | |
| XM_017008547.1 | 1 | 221 | XP_016864036.1 | |
| XM_017008548.1 | 1 | 221 | XP_016864037.1 | |
| XM_017008549.1 | 1 | 221 | XP_016864038.1 | |
| XM_017008550.1 | 1 | 221 | XP_016864039.1 | |
| XM_017008551.1 | 1 | 221 | XP_016864040.1 | |
| XM_017008552.1 | 1 | 221 | XP_016864041.1 | |
| XM_017008553.1 | 1 | 221 | XP_016864042.1 | |
| XM_017008554.1 | 1 | 221 | XP_016864043.1 | |
| XM_017008555.1 | 1 | 221 | XP_016864044.1 | |
| XM_017008556.1 | 1 | 221 | XP_016864045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830105 | Chr.4:145576238 - 145749385 on Build GRCh38 | Loss |
|

Genomic Map