Assay Details
Target Gene Details
Entrez Gene ID: | 27065 |
Gene Name: | neuron specific gene family member 1 |
Gene Aliases: |
D4S234, D4S234E, NEEP21, P21 |
Location: |
Chr.4:4380988-4419058 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSG1 | NM_001040101.1 | NP_001035190.1 | ||
| NM_001287763.1 | NP_001274692.1 | |||
| NM_001287764.1 | NP_001274693.1 | |||
| NM_014392.4 | NP_055207.1 | |||
| XM_017008022.1 | XP_016863511.1 | |||
| AK094573.1 | ||||
| AK125172.1 | ||||
| AK299657.1 | ||||
| AK301911.1 | ||||
| AK312288.1 | ||||
| AX775767.1 | CAE11583.1 | |||
| BC034227.1 | AAH34227.1 | |||
| DA746130.1 | ||||
| DA773692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436440 | Chr.4:4398087 - 4404887 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map