Assay Details
Target Gene Details
Entrez Gene ID: | 6452 |
Gene Name: | SH3 domain binding protein 2 |
Gene Aliases: |
3BP-2, 3BP2, CRBM, CRPM, RES4-23 |
Location: |
Chr.4:2793023-2841096 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3BP2 | NM_001122681.1 | NP_001116153.1 | ||
| AK225497.1 | ||||
| AK307106.1 | ||||
| AK312286.1 | ||||
| BC022996.1 | AAH22996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470002 | Chr.4:2699371 - 2834901 on Build GRCh38 | Loss |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv593435 | Chr.4:2769045 - 2879529 on Build GRCh38 | Loss |
|
| nsv516797 | Chr.4:2721804 - 2834901 on Build GRCh38 | Loss |
|
| nsv593436 | Chr.4:2792593 - 2845072 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map