Assay Details
Target Gene Details
Entrez Gene ID: | 152926 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1K |
Gene Aliases: |
BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07 |
Location: |
Chr.4:88257609-88284831 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPM1K | NM_152542.4 | NP_689755.3 | ||
XM_006714111.3 | XP_006714174.1 | |||
XM_017007803.1 | XP_016863292.1 | |||
XM_017007804.1 | XP_016863293.1 | |||
XM_017007805.1 | XP_016863294.1 | |||
XM_017007806.1 | XP_016863295.1 | |||
XM_017007807.1 | XP_016863296.1 | |||
AK054678.1 | BAB70790.1 | |||
AK314417.1 | ||||
AL834271.1 | CAD38946.1 | |||
AX766363.1 | CAE00325.1 | |||
AY157615.1 | AAO17296.1 | |||
AY435431.1 | AAR06213.1 | |||
AY994097.1 | AAX77016.1 | |||
BC037552.1 | AAH37552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759266 | Chr.4:87997399 - 88291967 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
esv3575758 | Chr.4:88250298 - 88318857 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
