Assay Details
Target Gene Details
Entrez Gene ID: | 201780 |
Gene Name: | solute carrier family 10 member 4 |
Gene Aliases: |
P4 |
Location: |
Chr.4:48483343-48489524 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A4 | NM_152679.3 | NP_689892.1 | ||
| AK126542.1 | ||||
| AY704414.1 | AAW30130.1 | |||
| BC012048.1 | AAH12048.2 | |||
| BC019066.2 | AAH19066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829925 | Chr.4:48465676 - 48637325 on Build GRCh38 | Gain+Loss |
|
| nsv594121 | Chr.4:48425756 - 48603611 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map