Assay Details
Target Gene Details
Entrez Gene ID: | 646576 |
Gene Name: | HHIP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:144642916-144646019 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHIP-AS1 | NR_037595.1 | 2 | 605 |

Genomic Map