Assay Details
Target Gene Details
Entrez Gene ID: | 27146 |
Gene Name: | family with sequence similarity 184 member B |
Gene Aliases: |
- |
Location: |
Chr.4:17632073-17781512 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM184B | NM_015688.1 | 17 | 3156 | NP_056503.1 |
| AB033102.1 | 18 | 2969 | ||
| AK027311.1 | 1 | 67 | ||
| AK308144.1 | 18 | 3157 |

Genomic Map