Assay Details
Target Gene Details
Entrez Gene ID: | 152877 |
Gene Name: | family with sequence similarity 53 member A |
Gene Aliases: |
DNTNP |
Location: |
Chr.4:1574055-1684313 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM53A | XM_011513404.2 | XP_011511706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8897n54 | Chr.4:1532249 - 1617803 on Build GRCh38 | Loss |
|
| nsv593316 | Chr.4:1526039 - 1768630 on Build GRCh38 | Gain |
|
| nsv949753 | Chr.4:1532074 - 1630173 on Build GRCh38 | Deletion |
|
| nsv593322 | Chr.4:1589655 - 1622987 on Build GRCh38 | Gain |
|
| nsv470000 | Chr.4:1522203 - 1722852 on Build GRCh38 | Loss |
|
| nsv593320 | Chr.4:1553021 - 1657401 on Build GRCh38 | Loss |
|
| nsv593321 | Chr.4:1584309 - 1601958 on Build GRCh38 | Gain |
|

Genomic Map