Assay Details
Target Gene Details
Entrez Gene ID: | 3093 |
Gene Name: | ubiquitin conjugating enzyme E2 K |
Gene Aliases: |
E2-25K, HIP2, HYPG, LIG, UBC1 |
Location: |
Chr.4:39698044-39782792 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2K | NM_001111112.1 | NP_001104582.1 | ||
| NM_001111113.1 | NP_001104583.1 | |||
| NM_001312646.1 | NP_001299575.1 | |||
| NM_001312647.1 | NP_001299576.1 | |||
| NM_001312648.1 | NP_001299577.1 | |||
| NM_005339.4 | NP_005330.1 | |||
| AB022435.1 | BAA78555.1 | |||
| AB022436.1 | BAA78556.1 | |||
| AK093533.1 | ||||
| AK291454.1 | ||||
| AK295846.1 | ||||
| AK307130.1 | ||||
| AK315524.1 | ||||
| BC022804.1 | AAH22804.1 | |||
| BC050600.1 | AAH50600.1 | |||
| CX786829.1 | ||||
| DA526283.1 | ||||
| U58522.1 | AAC50633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv441870 | Chr.4:39702789 - 39711713 on Build GRCh38 | Loss |
|
| nsv594026 | Chr.4:39708327 - 39712123 on Build GRCh38 | Loss |
|
| esv3563738 | Chr.4:39701381 - 39713206 on Build GRCh38 | Deletion |
|
| nsv518644 | Chr.4:39707188 - 39726889 on Build GRCh38 | Loss |
|
| nsv594029 | Chr.4:39708474 - 39714520 on Build GRCh38 | Loss |
|
| dgv1475e212 | Chr.4:39702601 - 39712707 on Build GRCh38 | Loss |
|
| esv2660659 | Chr.4:39702732 - 39712313 on Build GRCh38 | Deletion |
|
| nsv436937 | Chr.4:39692694 - 39736585 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map