Assay Details
Target Gene Details
Entrez Gene ID: | 27146 |
Gene Name: | family with sequence similarity 184 member B |
Gene Aliases: |
- |
Location: |
Chr.4:17632073-17781512 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM184B | NM_015688.1 | NP_056503.1 | ||
| AB033102.1 | ||||
| AK308144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv997752 | Chr.4:17643269 - 17648500 on Build GRCh38 | Deletion |
|
| nsv1006981 | Chr.4:17643727 - 17728912 on Build GRCh38 | Loss |
|

Genomic Map