Assay Details
Target Gene Details
Entrez Gene ID: | 55763 |
Gene Name: | exocyst complex component 1 |
Gene Aliases: |
BM-102, SEC3, SEC3L1, SEC3P |
Location: |
Chr.4:55853616-55905078 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOC1 | NM_001024924.1 | NP_001020095.1 | ||
| NM_018261.3 | NP_060731.2 | |||
| NM_178237.2 | NP_839955.1 | |||
| XM_005265747.2 | XP_005265804.1 | |||
| XM_005265748.1 | XP_005265805.1 | |||
| XM_005265750.2 | XP_005265807.1 | |||
| XM_017008405.1 | XP_016863894.1 | |||
| XM_017008406.1 | XP_016863895.1 | |||
| XM_017008407.1 | XP_016863896.1 | |||
| AK001755.1 | BAA91886.1 | |||
| AK027413.1 | BAB55095.1 | |||
| AL833208.1 | ||||
| BC094751.1 | AAH94751.1 | |||
| BI760313.1 | ||||
| BM468054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005845 | Chr.4:55655908 - 56114191 on Build GRCh38 | Gain |
|
| nsv10496 | Chr.4:55470779 - 55974971 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116059174] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map