Assay Details
Target Gene Details
Entrez Gene ID: | 84448 |
Gene Name: | actin binding LIM protein family member 2 |
Gene Aliases: |
- |
Location: |
Chr.4:7965310-8158832 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34140 | Chr.4:7786209 - 8170764 on Build GRCh38 | Loss |
|
| esv3599538 | Chr.4:7968979 - 7973413 on Build GRCh38 | Loss |
|
| esv23700 | Chr.4:7969526 - 7973205 on Build GRCh38 | Loss |
|
| nsv469779 | Chr.4:7963526 - 8175919 on Build GRCh38 | Loss |
|
| nsv829851 | Chr.4:7956971 - 8047333 on Build GRCh38 | Loss |
|
| esv2677688 | Chr.4:7968883 - 7973420 on Build GRCh38 | Deletion |
|
| esv3563370 | Chr.4:7968772 - 7973372 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map