Assay Details
Target Gene Details
Entrez Gene ID: | 26952 |
Gene Name: | submaxillary gland androgen regulated protein 3A |
Gene Aliases: |
P-B1, PBI, PRL5, PROL5 |
Location: |
Chr.4:70360776-70367158 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMR3A | NM_012390.3 | NP_036522.3 | ||
| AK312075.1 | ||||
| CD722466.1 | ||||
| CD722957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv187e55 | Chr.4:70360836 - 70386524 on Build GRCh38 | Loss |
|
| nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain |
|
| nsv1006332 | Chr.4:70295888 - 70412931 on Build GRCh38 | Loss |
|
| esv2759259 | Chr.4:70350067 - 70455756 on Build GRCh38 | Loss |
|
| nsv829964 | Chr.4:70274771 - 70439225 on Build GRCh38 | Gain |
|
| esv34954 | Chr.4:70360965 - 70371723 on Build GRCh38 | Loss |
|
| nsv461554 | Chr.4:70296786 - 70399455 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map