Assay Details
Target Gene Details
Entrez Gene ID: | 140458 |
Gene Name: | ankyrin repeat and SOCS box containing 5 |
Gene Aliases: |
ASB-5 |
Location: |
Chr.4:176213673-176277571 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB5 | NM_080874.3 | NP_543150.1 | ||
| XM_005262759.1 | XP_005262816.1 | |||
| XM_011531616.2 | XP_011529918.1 | |||
| XM_011531617.2 | XP_011529919.1 | |||
| AK096441.1 | BAC04791.1 | |||
| AK098693.1 | BAC05382.1 | |||
| AY057053.1 | AAL18248.1 | |||
| BC065710.1 | AAH65710.1 | |||
| BX647857.1 | CAI46074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020249 | Chr.4:176133684 - 176233735 on Build GRCh38 | Loss |
|
| nsv1017524 | Chr.4:176114932 - 176227598 on Build GRCh38 | Gain |
|
| esv34137 | Chr.4:175851071 - 176341258 on Build GRCh38 | Loss |
|
| esv2728772 | Chr.4:175817087 - 177875422 on Build GRCh38 | Deletion |
|
| esv3603274 | Chr.4:176144758 - 176386481 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map