Assay Details
Target Gene Details
Entrez Gene ID: | 132320 |
Gene Name: | sodium channel and clathrin linker 1 |
Gene Aliases: |
CAP-1A, CAP1A |
Location: |
Chr.4:128875206-129093607 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCLT1 | XM_017007717.1 | XP_016863206.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv9261n54 | Chr.4:128853220 - 129006649 on Build GRCh38 | Gain |
|
| esv3602167 | Chr.4:128849611 - 128942558 on Build GRCh38 | Gain |
|
| esv3575771 | Chr.4:128853799 - 129016711 on Build GRCh38 | Gain |
|
| dgv5386n100 | Chr.4:128849806 - 129043871 on Build GRCh38 | Gain |
|
| nsv517249 | Chr.4:128817173 - 129006649 on Build GRCh38 | Gain |
|
| esv3602168 | Chr.4:128851310 - 129020617 on Build GRCh38 | Gain |
|
| nsv4504 | Chr.4:128856213 - 128882804 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map