Assay Details
Target Gene Details
Entrez Gene ID: | 101929621 |
Gene Name: | uncharacterized LOC101929621 |
Gene Aliases: |
- |
Location: |
Chr.4:108557233-108560392 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929621 | NR_134679.1 |
Target Gene Details
Entrez Gene ID: | 285456 |
Gene Name: | RPL34 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.4:108538190-108620457 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL34-AS1 | NR_026968.1 | |||
| AK094992.1 | BAC04469.1 | |||
| AK298931.1 | ||||
| AK308624.1 |

Genomic Map