Assay Details
Target Gene Details
Entrez Gene ID: | 26049 |
Gene Name: | family with sequence similarity 169 member A |
Gene Aliases: |
SLAP75 |
Location: |
Chr.5:74777574-74867994 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM169A | NM_015566.2 | NP_056381.1 | ||
| NR_046462.1 | ||||
| XM_005248480.2 | XP_005248537.1 | |||
| XM_011543306.2 | XP_011541608.1 | |||
| XM_011543307.2 | XP_011541609.1 | |||
| XM_017009332.1 | XP_016864821.1 | |||
| XM_017009333.1 | XP_016864822.1 | |||
| AB020695.1 | ||||
| AK022981.1 | BAB14344.1 | |||
| AK290004.1 | ||||
| BX640902.1 | CAE45948.1 | |||
| BX647536.1 | ||||
| CR936791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv598653 | Chr.5:74697056 - 74799717 on Build GRCh38 | Loss |
|

Genomic Map