Assay Details
Target Gene Details
Entrez Gene ID: | 651746 |
Gene Name: | ankyrin repeat domain 33B |
Gene Aliases: |
- |
Location: |
Chr.5:10564323-10657816 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD33B | NM_001164440.1 | NP_001157912.1 | ||
| XM_005248344.3 | XP_005248401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597125 | Chr.5:10577783 - 10611467 on Build GRCh38 | Loss |
|

Genomic Map