Assay Details
Target Gene Details
Entrez Gene ID: | 54540 |
Gene Name: | family with sequence similarity 193 member B |
Gene Aliases: |
IRIZIO |
Location: |
Chr.5:177519789-177554585 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5148 | Chr.5:177519430 - 177552310 on Build GRCh38 | Insertion |
|
| nsv5147 | Chr.5:177511195 - 177556017 on Build GRCh38 | Deletion |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|

Genomic Map