Assay Details
Target Gene Details
Entrez Gene ID: | 26049 |
Gene Name: | family with sequence similarity 169 member A |
Gene Aliases: |
SLAP75 |
Location: |
Chr.5:74777574-74867994 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM169A | NM_015566.2 | 10 | 1088 | NP_056381.1 |
| NR_046462.1 | 9 | 1051 | ||
| XM_005248480.2 | 10 | 1223 | XP_005248537.1 | |
| XM_011543306.2 | 10 | 1310 | XP_011541608.1 | |
| XM_011543307.2 | 10 | 1739 | XP_011541609.1 | |
| XM_017009332.1 | 8 | 1986 | XP_016864821.1 | |
| XM_017009333.1 | 8 | 960 | XP_016864822.1 | |
| AB020695.1 | 10 | 1040 | ||
| AK022981.1 | 1 | 44 | BAB14344.1 | |
| AK290004.1 | 10 | 1088 | ||
| BX640902.1 | 9 | 869 | CAE45948.1 | |
| BX647536.1 | 9 | 1053 | ||
| CR936791.1 | 10 | 1090 |

Genomic Map