Assay Details
Target Gene Details
Entrez Gene ID: | 22993 |
Gene Name: | HMG-box containing 3 |
Gene Aliases: |
HMGX3, SMF |
Location: |
Chr.5:150000606-150053143 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGXB3 | NM_014983.2 | NP_055798.2 | ||
| XM_006714765.2 | XP_006714828.1 | |||
| BC051025.1 | AAH51025.1 | |||
| BC092520.1 | AAH92520.1 | |||
| D83778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv24664 | Chr.5:149854140 - 150050084 on Build GRCh38 | Gain+Loss |
|
| nsv830518 | Chr.5:149940811 - 150117140 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map