Assay Details
Target Gene Details
Entrez Gene ID: | 23176 |
Gene Name: | septin 8 |
Gene Aliases: |
SEP2 |
Location: |
Chr.5:132750817-132778216 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPT8 | NM_001098811.1 | 5 | 849 | NP_001092281.1 |
| NM_001098812.1 | 5 | 849 | NP_001092282.1 | |
| NM_001098813.1 | 5 | 775 | NP_001092283.1 | |
| NM_001300798.1 | 5 | 843 | NP_001287727.1 | |
| NM_001300799.1 | 5 | 843 | NP_001287728.1 | |
| NM_015146.1 | 5 | 849 | NP_055961.1 | |
| XM_005271927.2 | 5 | 851 | XP_005271984.1 | |
| XM_005271928.2 | 5 | 845 | XP_005271985.1 | |
| XM_005271931.3 | 6 | 932 | XP_005271988.1 | |
| XM_011543288.1 | 5 | 871 | XP_011541590.1 | |
| XM_017009254.1 | 5 | 845 | XP_016864743.1 | |
| XM_017009255.1 | 5 | 918 | XP_016864744.1 | |
| XM_017009256.1 | 5 | 912 | XP_016864745.1 | |
| XM_017009257.1 | 5 | 865 | XP_016864746.1 | |
| XM_017009258.1 | 5 | 918 | XP_016864747.1 | |
| XM_017009259.1 | 5 | 912 | XP_016864748.1 | |
| XM_017009260.1 | 5 | 865 | XP_016864749.1 | |
| XM_017009261.1 | 5 | 912 | XP_016864750.1 | |
| AF179995.1 | 5 | 849 | AAG09407.1 | |
| AF440761.1 | 5 | 849 | AAO13878.1 | |
| AF440762.1 | 5 | 849 | AAO13879.1 | |
| AF440763.1 | 5 | 684 | AAO13880.1 | |
| AK057797.1 | 5 | 707 | ||
| BC001329.1 | 2 | 248 | AAH01329.1 | |
| BC140759.1 | 5 | 665 | ||
| BC171779.1 | 5 | 665 | ||
| D86957.1 | 5 | 849 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28018 | Chr.5:132759610 - 132763721 on Build GRCh38 | Gain |
|
| nsv528687 | Chr.5:132750285 - 132779155 on Build GRCh38 | Gain |
|
| esv3606809 | Chr.5:132749939 - 132764024 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map