Assay Details
Target Gene Details
Entrez Gene ID: | 3340 |
Gene Name: | N-deacetylase and N-sulfotransferase 1 |
Gene Aliases: |
HSST, MRT46, NST1 |
Location: |
Chr.5:150485783-150558211 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDST1 | XM_005268433.1 | XP_005268490.1 | ||
| XM_005268435.4 | XP_005268492.1 | |||
| XM_005268437.4 | XP_005268494.1 | |||
| XM_005268438.1 | XP_005268495.1 | |||
| XM_005268439.1 | XP_005268496.1 | |||
| XM_005268441.1 | XP_005268498.1 | |||
| XM_006714783.1 | XP_006714846.1 | |||
| XM_011537638.2 | XP_011535940.1 | |||
| XM_017009427.1 | XP_016864916.1 | |||
| XM_017009428.1 | XP_016864917.1 | |||
| XM_017009429.1 | XP_016864918.1 | |||
| AB209107.1 | BAD92344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
|
| nsv462488 | Chr.5:150477897 - 150576147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map