Assay Details
Target Gene Details
Entrez Gene ID: | 54508 |
Gene Name: | EPB41L4A antisense RNA 2 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.5:112419583-112420978 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPB41L4A-AS2 | NR_027706.1 | |||
| AK002097.1 | BAA92083.1 | |||
| BC107117.2 | ||||
| BC107118.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950114 | Chr.5:112418904 - 112420403 on Build GRCh38 | Deletion |
|
| nsv599368 | Chr.5:112064188 - 112696978 on Build GRCh38 | Gain |
|

Genomic Map