Assay Details
Target Gene Details
Entrez Gene ID: | 728780 |
Gene Name: | ankyrin repeat and death domain containing 1B |
Gene Aliases: |
- |
Location: |
Chr.5:75611289-75671846 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKDD1B | NM_001276713.1 | NP_001263642.1 | ||
| XM_011543618.2 | 1 | 130 | XP_011541920.1 | |
| XM_011543623.2 | 1 | 130 | XP_011541925.1 | |
| XM_017009814.1 | 1 | 130 | XP_016865303.1 | |
| XM_017009815.1 | 1 | 130 | XP_016865304.1 | |
| XM_017009816.1 | 1 | 130 | XP_016865305.1 | |
| XM_017009817.1 | 1 | 130 | XP_016865306.1 | |
| AK307154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518475 | Chr.5:75567216 - 76115979 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map