Assay Details
Target Gene Details
Entrez Gene ID: | 728264 |
Gene Name: | cardiac mesoderm enhancer-associated non-coding RNA |
Gene Aliases: |
CARMEN, MIR143HG |
Location: |
Chr.5:149406845-149432836 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARMN | NR_105059.1 | |||
| NR_105060.1 | ||||
| AK126481.1 | ||||
| BX640700.1 | 2 | 4005 | ||
| CR933654.1 | ||||
| HQ848934.1 | ||||
| HQ848935.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521406 | Chr.5:149405886 - 149423248 on Build GRCh38 | Loss |
|
| nsv519878 | Chr.5:149408099 - 149419169 on Build GRCh38 | Loss |
|
| nsv599948 | Chr.5:149346599 - 149413198 on Build GRCh38 | Loss |
|

Genomic Map