Assay Details
Target Gene Details
| Entrez Gene ID: | 10923 | 
| Gene Name: | SUB1 homolog, transcriptional regulator | 
| Gene Aliases: | P15, PC4, p14 | 
| Location: | Chr.5:32531778-32604079 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| SUB1 | XM_017008986.1 | XP_016864475.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2759338 | Chr.5:32245255 - 32738538 on Build GRCh38 | Loss |  ZFR  MIR579  MTMR12  SUB1  NPR3 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top