Assay Details
Target Gene Details
Entrez Gene ID: | 54540 |
Gene Name: | family with sequence similarity 193 member B |
Gene Aliases: |
IRIZIO |
Location: |
Chr.5:177519789-177554585 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM193B | NM_001190946.2 | 6 | 2047 | NP_001177875.1 |
| NR_024019.3 | 9 | 3706 | ||
| XM_006714875.2 | 7 | 2175 | XP_006714938.1 | |
| XM_006714876.1 | 7 | 2898 | XP_006714939.1 | |
| XM_006714878.1 | 8 | 2752 | XP_006714941.1 | |
| XM_006714879.3 | 8 | 1995 | XP_006714942.1 | |
| XM_006714880.1 | 7 | 2793 | XP_006714943.1 | |
| XM_006714884.1 | 5 | 1296 | XP_006714947.1 | |
| XM_006714885.1 | 4 | 1100 | XP_006714948.1 | |
| XM_011534572.1 | 7 | 2070 | XP_011532874.1 | |
| XM_011534573.1 | 8 | 2170 | XP_011532875.1 | |
| XM_011534574.1 | 8 | 2615 | XP_011532876.1 | |
| XM_011534579.2 | 7 | 1934 | XP_011532881.1 | |
| XM_011534580.1 | 8 | 2959 | XP_011532882.1 | |
| XM_011534581.1 | 7 | 2639 | XP_011532883.1 | |
| XM_011534582.1 | 8 | 3146 | XP_011532884.1 | |
| XM_011534583.1 | 8 | 2887 | XP_011532885.1 | |
| AB067518.1 | 9 | 3663 | BAB67824.1 | |
| AK001266.1 | 3 | 976 | BAA91589.1 | |
| AK096032.1 | 7 | 1858 | ||
| AL833096.1 | 1 | 927 | ||
| BC008784.2 | 5 | 1296 | AAH08784.1 | |
| BC071696.1 | 3 | 950 | AAH71696.1 | |
| BX647587.1 | 6 | 1791 | CAI46078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5148 | Chr.5:177519430 - 177552310 on Build GRCh38 | Insertion |
|
| nsv5147 | Chr.5:177511195 - 177556017 on Build GRCh38 | Deletion |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|

Genomic Map