Assay Details
Target Gene Details
Entrez Gene ID: | 4245 |
Gene Name: | mannosyl (alpha-1,3-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase |
Gene Aliases: |
GLCNAC-TI, GLCT1, GLYT1, GNT-1, GNT-I, GnTI, MGAT |
Location: |
Chr.5:180790541-180815652 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT1 | NM_001114617.1 | NP_001108089.1 | ||
| NM_001114618.1 | NP_001108090.1 | |||
| XM_005265915.1 | XP_005265972.1 | |||
| XM_005265916.1 | XP_005265973.1 | |||
| XM_006714866.1 | XP_006714929.1 | |||
| XM_011534559.1 | XP_011532861.1 | |||
| XM_011534560.1 | XP_011532862.1 | |||
| XM_011534561.1 | XP_011532863.1 | |||
| XM_011534562.1 | XP_011532864.1 | |||
| XM_011534563.2 | XP_011532865.1 | |||
| XM_017009486.1 | XP_016864975.1 | |||
| XM_017009487.1 | XP_016864976.1 | |||
| AK057194.1 | ||||
| AK092256.1 | ||||
| AK094130.1 | ||||
| BG337235.1 | ||||
| DB270226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161322 | Chr.5:180717688 - 180914895 on Build GRCh38 | Duplication |
|
| nsv428133 | Chr.5:180805451 - 181359487 on Build GRCh38 | Gain |
|
| dgv9n29 | Chr.5:180758452 - 180976232 on Build GRCh38 | Loss |
|
| esv2759399 | Chr.5:180652970 - 180854476 on Build GRCh38 | Gain |
|
| nsv1026417 | Chr.5:180294376 - 180864360 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77429283] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map