Assay Details
Target Gene Details
Entrez Gene ID: | 728264 |
Gene Name: | cardiac mesoderm enhancer-associated non-coding RNA |
Gene Aliases: |
CARMEN, MIR143HG |
Location: |
Chr.5:149406845-149432836 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARMN | NR_105059.1 | |||
| NR_105060.1 | ||||
| AK126481.1 | 5 | 2082 | ||
| AL137734.1 | 1 | 2419 | ||
| CR933654.1 | 2 | 5048 | ||
| HQ848934.1 | ||||
| HQ848935.1 |

Genomic Map