Assay Details
Target Gene Details
Entrez Gene ID: | 23379 |
Gene Name: | interactor of little elongation complex ELL subunit 1 |
Gene Aliases: |
KIAA0947 |
Location: |
Chr.5:5422673-5490234 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ICE1 | NM_015325.2 | NP_056140.1 | ||
| XM_011513999.2 | XP_011512301.1 | |||
| XM_017009285.1 | XP_016864774.1 | |||
| AB023164.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021802 | Chr.5:5415764 - 5452773 on Build GRCh38 | Gain+Loss |
|
| esv3603900 | Chr.5:5400618 - 5426975 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map