Assay Details
Target Gene Details
Entrez Gene ID: | 57688 |
Gene Name: | zinc finger SWIM-type containing 6 |
Gene Aliases: |
AFND |
Location: |
Chr.5:61332273-61546172 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSWIM6 | NM_020928.1 | NP_065979.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3605213 | Chr.5:61335081 - 61344791 on Build GRCh38 | Loss |
|
| nsv950069 | Chr.5:61333374 - 61347473 on Build GRCh38 | Duplication |
|
| nsv598319 | Chr.5:61331130 - 61341001 on Build GRCh38 | Loss |
|
| nsv823089 | Chr.5:61293674 - 61374076 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80200332] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map