Assay Details
Target Gene Details
Entrez Gene ID: | 256006 |
Gene Name: | ankyrin repeat domain 31 |
Gene Aliases: |
- |
Location: |
Chr.5:75068297-75236897 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD31 | NM_001164443.1 | NP_001157915.1 | ||
| XM_011543298.2 | XP_011541600.1 | |||
| XM_011543299.1 | XP_011541601.1 | |||
| XM_011543300.2 | XP_011541602.1 | |||
| XM_011543301.2 | XP_011541603.1 | |||
| XM_011543302.2 | XP_011541604.1 | |||
| XM_017009317.1 | XP_016864806.1 | |||
| XM_017009318.1 | XP_016864807.1 | |||
| XM_017009319.1 | XP_016864808.1 | |||
| XM_017009320.1 | XP_016864809.1 | |||
| XM_017009321.1 | XP_016864810.1 |

Genomic Map