Assay Details
Target Gene Details
Entrez Gene ID: | 3101 |
Gene Name: | hexokinase 3 |
Gene Aliases: |
HKIII, HXK3 |
Location: |
Chr.5:176880869-176899373 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HK3 | NM_002115.2 | NP_002106.2 | ||
| XM_017009411.1 | XP_016864900.1 | |||
| AB208951.1 | BAD92188.1 | |||
| BC028129.1 | AAH28129.1 | |||
| U42303.1 | AAC50422.1 | |||
| U51333.1 | AAC50732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950724 | Chr.5:176766400 - 176890599 on Build GRCh38 | Deletion |
|
| dgv10181n54 | Chr.5:176808074 - 176883197 on Build GRCh38 | Loss |
|
| nsv462562 | Chr.5:176836148 - 176883197 on Build GRCh38 | Loss |
|
| dgv10180n54 | Chr.5:176741506 - 176883197 on Build GRCh38 | Loss |
|
| dgv362n21 | Chr.5:176878677 - 176889200 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map