Assay Details
Target Gene Details
Entrez Gene ID: | 153469 |
Gene Name: | JAKMIP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:147559994-147662009 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAKMIP2-AS1 | NR_038902.1 | |||
| AK027578.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2730891 | Chr.5:147561178 - 147561607 on Build GRCh38 | Deletion |
|
| esv1978895 | Chr.5:147561191 - 147561628 on Build GRCh38 | Deletion |
|
| esv2730890 | Chr.5:147560941 - 147561968 on Build GRCh38 | Deletion |
|

Genomic Map