Assay Details
Target Gene Details
Entrez Gene ID: | 9832 |
Gene Name: | janus kinase and microtubule interacting protein 2 |
Gene Aliases: |
JAMIP2, NECC1 |
Location: |
Chr.5:147585439-147782848 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAKMIP2 | NM_001270934.1 | 21 | 8454 | NP_001257863.1 |
| NM_001270941.1 | 22 | 8517 | NP_001257870.1 | |
| NM_001282282.1 | 21 | 8240 | NP_001269211.1 | |
| NM_014790.4 | 21 | 8446 | NP_055605.2 | |
| XM_017010097.1 | 23 | 8511 | XP_016865586.1 | |
| XM_017010098.1 | 21 | 8397 | XP_016865587.1 |
Target Gene Details
Entrez Gene ID: | 153469 |
Gene Name: | JAKMIP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:147559994-147662009 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAKMIP2-AS1 | NR_038902.1 | |||
| AK027578.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78381583,rs79451119] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map