Assay Details
Target Gene Details
Entrez Gene ID: | 2908 |
Gene Name: | nuclear receptor subfamily 3 group C member 1 |
Gene Aliases: |
GCCR, GCR, GCRST, GR, GRL |
Location: |
Chr.5:143277931-143435512 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5041 | Chr.5:143273624 - 143318331 on Build GRCh38 | Deletion |
|
| esv33748 | Chr.5:143258702 - 143320030 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148700478] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map