Assay Details
Target Gene Details
Entrez Gene ID: | 23176 |
Gene Name: | septin 8 |
Gene Aliases: |
SEP2 |
Location: |
Chr.5:132750817-132778216 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890740 | Chr.5:132767579 - 132823442 on Build GRCh38 | Loss |
|
| nsv528687 | Chr.5:132750285 - 132779155 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map