Assay Details
Target Gene Details
Entrez Gene ID: | 51663 |
Gene Name: | zinc finger RNA binding protein |
Gene Aliases: |
SPG71, ZFR1 |
Location: |
Chr.5:32354350-32444738 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFR | NM_016107.3 | NP_057191.2 | ||
| AF100742.1 | AAD40385.1 | |||
| AJ314790.1 | CAC40818.1 | |||
| AK025481.1 | BAB15147.1 | |||
| AK055938.1 | ||||
| AK123306.1 | ||||
| AK295297.1 | ||||
| AK307808.1 | ||||
| AL137258.1 | CAB70659.1 | |||
| BC051893.1 | AAH51893.1 | |||
| BC062986.1 | AAH62986.1 | |||
| BC137084.1 | ||||
| BC137085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021781 | Chr.5:32102613 - 32412387 on Build GRCh38 | Gain |
|
| esv2730025 | Chr.5:32356046 - 32364166 on Build GRCh38 | Deletion |
|
| esv2759338 | Chr.5:32245255 - 32738538 on Build GRCh38 | Loss |
|
| esv3604608 | Chr.5:32357219 - 32363950 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74571056] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map