Assay Details
Target Gene Details
Entrez Gene ID: | 54545 |
Gene Name: | myotubularin related protein 12 |
Gene Aliases: |
3-PAP, PIP3AP |
Location: |
Chr.5:32227007-32313008 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021781 | Chr.5:32102613 - 32412387 on Build GRCh38 | Gain |
|
| dgv203e55 | Chr.5:32114037 - 32260077 on Build GRCh38 | Gain |
|
| esv3604580 | Chr.5:31525629 - 32360060 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map