Assay Details
Target Gene Details
Entrez Gene ID: | 100129716 |
Gene Name: | ARRDC3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:91380347-91420715 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC3-AS1 | NR_027435.1 |

Genomic Map