Assay Details
Target Gene Details
Entrez Gene ID: | 23197 |
Gene Name: | Fas associated factor family member 2 |
Gene Aliases: |
ETEA, UBXD8, UBXN3B |
Location: |
Chr.5:176448333-176510074 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAF2 | NM_014613.2 | NP_055428.1 | ||
| XM_011534475.2 | XP_011532777.1 | |||
| AB020694.1 | BAA74910.1 | |||
| AB088120.1 | BAC22491.1 | |||
| AK304346.1 | ||||
| AL706430.1 | ||||
| BC001069.1 | ||||
| BC006145.1 | ||||
| BC014001.2 | AAH14001.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023853 | Chr.5:176043309 - 176463537 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79208532] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map