Assay Details
Target Gene Details
Entrez Gene ID: | 23120 |
Gene Name: | ATPase phospholipid transporting 10B (putative) |
Gene Aliases: |
ATPVB |
Location: |
Chr.5:160563120-160929411 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP10B | NM_025153.2 | NP_079429.2 | ||
| XM_006714833.2 | XP_006714896.1 | |||
| XM_011534468.2 | XP_011532770.1 | |||
| XM_011534469.1 | XP_011532771.1 | |||
| XM_011534472.2 | XP_011532774.1 | |||
| XM_017009252.1 | XP_016864741.1 | |||
| XM_017009253.1 | XP_016864742.1 | |||
| AB018258.2 | ||||
| AK025290.1 | ||||
| AK025294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026980 | Chr.5:160438325 - 160798037 on Build GRCh38 | Gain |
|
| nsv1021499 | Chr.5:160531332 - 160735337 on Build GRCh38 | Gain |
|
| nsv1015634 | Chr.5:159750943 - 160620377 on Build GRCh38 | Gain |
|
| esv3607372 | Chr.5:160433056 - 160568517 on Build GRCh38 | Gain |
|
| esv3607375 | Chr.5:160558982 - 160607722 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map